Learn More
Ansell™ Latex Canners and Handlers Gloves
Unlined industrial latex gloves
Manufacturer:  Ansell™ 88392 8.5 193924
 View more versions of this product
Catalog No. 19-014575
  Check Availability
Add to cart
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
Pebble embossed
canning and food processing, small parts assembly, pharmaceutical manufacturing and plant maintenance, resist Acids, Detergents, vegetable Oil, Salts, Caustics and Alcohols
8.5
21 CFR 177-2600, FDA
Natural
30.5cm
Pinked
Rubber, natural
20 mil
Powder-free
12 in.