Ansell™ Neox™ Supported Neoprene Gloves
Reinforced neoprene coating provides exceptional resistance to abrasion, cutting, and chemicals
Manufacturer: Ansell™ 9912 10212986
 View more versions of this product
Catalog No. 19-014955
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Certifications 21 CFR 177-2600
    Glove Cuff Style Gauntlet
    For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, handling Oils, Acids, Caustics and Alcohols, handling specialty chemicals, hazmat work, plating operations
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Lining Two-piece Fleece, Jersey knit
    Material Neoprene
    Size 10
    Coating Material Smooth
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback