Ansell™ Omni™ Gloves
Cotton flock lined for maximum user comfort
Manufacturer: Ansell™ 00276 9 192765
 View more versions of this product
Catalog No. 11-394-90C
  Check Availability
Add to Cart
Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
    Specifications
    Certifications 21 CFR 177-2600, FDA
    For Use With (Application) blend of Neoprene and natural Latex resists tearing, puncturing, abrasions and various chemicals, including Caustics, animal fats, Alcohols, Acids, Salts, Detergents and Petroleum based Solvents
    Glove Cuff Style Embossed Pebble
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Lining Cotton Flock
    Material Neoprene, Natural Rubber Latex
    Size 9
    Thickness 20mil
    Color Light green
  • Description & Specifications
Related Products
Product Content Feedback