Ansell™ Ansell™ Overhead-Style Vinyl Coat Aprons with Open Back
Cooler to wear in situations requiring front splash protection only
Manufacturer: Ansell™ 56 902 L 950166
 View more versions of this product
Catalog No. 17-987-124C
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Dimensions (L x W) 54 x 56 in. (137.16 x 142.2cm)
    Cuff Style Elastic cuffs
    Material Vinyl
    Color Blue
    Size Large
    Aprons feature an open back that leaves approximately 6 to 8 in. (15.2 to 20.3cm) of the wearer's back exposed.
  • Description & Specifications
Related Products
Product Content Feedback