Learn More
Ansell™ Polar Bear™ Plus 35 Gloves
Made with high-performance Spectra™ fibers
Manufacturer:  Ansell™ 74035 9 222067
 View more versions of this product
Catalog No. 19-058964
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Lightweight, soft, and exceptionally flexible. Ideal for use in meatpacking, food processing, materials handling and numerous other applications requiring an exceptionally high level of protection from cuts.
  • Combines high-performance fiber with other ultratough filaments
  • Provides outstanding protection from cuts and abrasion
  • Can be worn comfortably for extended periods
  • Machine launderable and hand-to-hand reversible, so no pairing is required after laundering
  • Rib-knitted wrist and high-visibility color-coded wrist trim
  • Color: White
Order Info
Sold by the glove.
Specifications
Specifications
Safety
Tuff-Cuff™
HDPE
Heavy weight
White
Level 5
beef, pork, poultry, fish processing, canning and food processing, cleaning and changing stationary slicer blades in delis and supermarkets
9
White