Learn More
Ansell™ Polar Bear™ Plus 35 Gloves
Made with high-performance Spectra™ fibers
Manufacturer:  Ansell™ 222065
 View more versions of this product
Catalog No. 19-058965
Add to cart
Description
Description
Lightweight, soft, and exceptionally flexible. Ideal for use in meatpacking, food processing, materials handling and numerous other applications requiring an exceptionally high level of protection from cuts.
  • Combines high-performance fiber with other ultratough filaments
  • Provides outstanding protection from cuts and abrasion
  • Can be worn comfortably for extended periods
  • Machine launderable and hand-to-hand reversible, so no pairing is required after laundering
  • Rib-knitted wrist and high-visibility color-coded wrist trim
  • Color: White
Order Info
Sold by the glove.
Specifications
Specifications
Safety
Tuff-Cuff™
HDPE
Heavy weight
White
Level 5
beef, pork, poultry, fish processing, canning and food processing, cleaning and changing stationary slicer blades in delis and supermarkets
7
White