Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves
Click to view available options
Size:
8
9
10
11
Description
- Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
- Neoprene foam coating enhances grip in dry, wet, or oily conditions
- Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
- Ergonomic design is comfortable and flexible
Specifications
ANSI Abrasion Rating | ANSI 3 |
Arc Flash | Level 2 |
Certifications/Compliance | NFPA 70E Hazard Risk Category 2 |
Certifications/Compliance (Europe) | EN 407: 412110 |
Coating Material | Neoprene |
Color | Green, Black |
Cut Resistance Rating | ANSI A4 |
Hand | Specific |
Material | Aramid Fiber |
Size | 10 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction