Learn More
Ansell PowerFlex Plus Gloves
Well suited for sheet metal and glass handling, recycling, canning, and bottling applications
$92.00
Specifications
Exterior Finish | Textured |
---|---|
For Use With (Application) | sheet Metal and Glass handling, recycling, canning and bottling applications |
Glove Cuff Style | Knit |
Liner Material | Dupont™ Kevlar™ aramid fibers |
Color | Blue, Yellow |
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-170-504C
|
Ansell
806008 |
8 |
Dozen of 12 for $92.00
Case of 6 DZ for $552.00 |
|
19-133-487
|
Ansell
80600 10206411 |
10 |
Dozen of 12 for $92.00
Case of 6 DZ for $552.00 |
|
19-170-504D
|
Ansell
806009 |
9 |
Dozen of 12 for $92.00
Case of 6 DZ for $552.00 |
|
Description
Seamless 100% Kevlar cut-resistant knit lining and textured natural rubber coating on palm and full thumb provide excellent cut, abrasion, and puncture resistance.
- Exceptional wet and dry grip
- Uncoated back allows air circulation to keep hands cooler, drier
- Yellow with blue coating
Specifications
Textured | |
Knit | |
Blue, Yellow |
sheet Metal and Glass handling, recycling, canning and bottling applications | |
Dupont™ Kevlar™ aramid fibers | |
Rubber |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.