Ansell™ Redmont™ Neoprene Gloves

Ideal for applications involving petrochemicals and refining, chemical transport, and hazardous materials

Manufacturer: Ansell™ 19938 10 215026

 View more versions of this product

Catalog No. 19-806-040

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Gauntlet
    For Use With (Application) Petrochemicals and refining, chemical transport and hazmat work
    Length (English) 18 in.
    Length (Metric) 45.72cm
    Lining Fleece, Jersey
    Material Neoprene
    Size 10
    Coating Material Rough
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit