Learn More
Ansell™ Sawyer-Tower™ 66-677 Series Coveralls
Full-coverage chemical protection with added safety of fire resistance
Manufacturer:  Ansell™ 66677L
 View more versions of this product
Catalog No. 19-010-047
Add to cart
This item is not returnable. View return policy
Description
Description
  • Features comfortable Gore™ Chemical Splash Fabric
  • Chemical splash protection
  • Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
  • CPC Nomex provides fire-resistant chemical splash protection
  • Special sizes may be available upon request
  • Flame resistance
  • HRC rating: 2
Specifications
Specifications
CPC Nomex™ Tri-laminate
Blue
Large
2
Flame, Chemical