Learn More
Ansell™ Sawyer-Tower™ 66-677 Series Coveralls
Full-coverage chemical protection with added safety of fire resistance
$1070.19 - $1326.86
Specifications
Material CPC Nomex™ Tri-laminate
Color Blue
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-010-046
View Documents
Ansell™
66677M
Medium
Each for $1,070.19
Case of 5 EA for $5,350.93
Add to cart
19-010-047
View Documents
Ansell™
66677L
Large
Each for $1,112.62
Case of 5 EA for $5,563.10
Add to cart
19-010-048
View Documents
Ansell™
66677XL
X-Large
Each for $1,151.95
Case of 5 EA for $5,759.76
Add to cart
19-010-049
View Documents
Ansell™
666772XL
2X-Large
Each for $1,326.86
Case of 5 EA for $6,634.31
Add to cart
Description
Description
  • Features comfortable Gore™ Chemical Splash Fabric
  • Chemical splash protection
  • Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
  • CPC Nomex provides fire-resistant chemical splash protection
  • Special sizes may be available upon request
  • Flame resistance
  • HRC rating: 2
Specifications
Specifications
CPC Nomex™ Tri-laminate
Blue