Learn More
Ansell™ AlphaTec™ 39-124 Nitrile Gloves
Provides heavy-duty mechanical and chemical protection for wide variety of work environments. Sol-Knit™ Nitrile Heavy Duty Gloves have better mechanical performance and chemical protection.
Manufacturer:  Ansell™ 217806
 View more versions of this product
Catalog No. 19-806-053
Add to cart
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
  • Liquid and chemical protection gloves
  • Industrial gloves
  • Tough glove offers optimum comfort of use thanks to its natural shape that makes it well suited for use with long-lasting tasks
  • Superior dexterity and the high levels of wet and dry grip contribute to wearer safety and ultimately to the overall productivity
  • Rugged glove with chemical protection
Applications:
  • Adjusting panels parts components
  • Applying finish to materials, products
  • Assembly and disassembly of equipment
  • Equipment repair, maintenance
  • Handling materials with sharp or rough edges
  • Oil, fluids and filter change
  • Production line support and maintenance
Compliance
Meets ANSI A1 (cut level) and ANSI 4 (abrasion level); EN 374, EN 388, EN 407; FDA 510(k) approved.
Specifications
Specifications
Safety
Gauntlet
Yes
Nitrile
Interlock cotton
9
Nitrile gloves
14 in.
21 CFR 177-2600, FDA, EN 420:2003+A1:2009 and EN 388:2003 , EN 407:2004 , EN 374:2003
Rough
chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Cotton
12 pairs in a bag, 6 bags in a carton
39-124
Supported
Green
350 mm