Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ AlphaTec™ 39-124 Nitrile Gloves

Provides heavy-duty mechanical and chemical protection for wide variety of work environments. Sol-Knit™ Nitrile Heavy Duty Gloves have better mechanical performance and chemical protection.

$189.10

Specifications

Color Green
Material Nitrile
Glove Cuff Style Gauntlet
View More Specs

Alert:

Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.


Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-806-114 View Documents Ansell™
217804
7 Pack of 12 for $189.10
Case of 6 PK for $1,168.00
Add to cart
 
19-806-115 View Documents Ansell™
39124/8
8 Pack of 12 for $189.10
Case of 6 PK for $1,168.00
Add to cart
 
19-806-116 View Documents Ansell™
39124/9
9 Pack of 12 for $189.10
Case of 6 PK for $1,168.00
Add to cart
 
19-806-053 View Documents Ansell™
217806
9 Dozen of 12 for $189.10
Case of 6 DZ for $1,168.00
Add to cart
 
19-806-117 View Documents Ansell™
39124/10
10 Pack of 12 for $189.10
Case of 6 PK for $1,168.00
Add to cart
 
Description

Description

  • Liquid and chemical protection gloves
  • Industrial gloves
  • Tough glove offers optimum comfort of use thanks to its natural shape that makes it well suited for use with long-lasting tasks
  • Superior dexterity and the high levels of wet and dry grip contribute to wearer safety and ultimately to the overall productivity
  • Rugged glove with chemical protection

Applications:

  • Adjusting panels parts components
  • Applying finish to materials, products
  • Assembly and disassembly of equipment
  • Equipment repair, maintenance
  • Handling materials with sharp or rough edges
  • Oil, fluids and filter change
  • Production line support and maintenance

Compliance

Meets ANSI A1 (cut level) and ANSI 4 (abrasion level); EN 374, EN 388, EN 407; FDA 510(k) approved.

Specifications

Specifications

Green
Nitrile
Gauntlet
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit