Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell Sol-Knit Gloves
Offers chemical resistance of nitrile with the strength and comfort of cotton
$76.00
Specifications
Certifications/Compliance | 21 CFR 177-2600, FDA |
---|---|
Color | Green |
Length (English) | 14 in. |
Length (Metric) | 35.5 cm |
Material | Nitrile |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-120-394D
|
Ansell
217807 |
10 |
Dozen of 12 for $76.00
Case of 6 DZ for $456.00 |
|
Description
- Gauntlet style
- Length: 14 in.
Certifications
21 CFR 177-2600, FDA.
Specifications
21 CFR 177-2600, FDA | |
14 in. | |
Nitrile | |
Gauntlet |
Green | |
35.5 cm | |
Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work | |
Cotton |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title