Ansell™ Sol-Vex™ Nitrile Gloves
Resistant to chemicals—will not weaken or swell in many solvents, caustics or animal fats. Ansell™ Sol-Vex™ Nitrile Gloves are ideal for numerous applications involving chemical processing.
Manufacturer: Ansell™ 37155 11 117145
 View more versions of this product
Catalog No. 11-391-137E
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Glove Cuff Style Straight
    For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
    Length (English) 13 in.
    Length (Metric) 33cm
    Lining None
    Material Nitrile
    Palm Type Contoured
    Size 11
    Thickness 15mil
    Color Green
  • Description & Specifications
Related Products
Product Content Feedback