Ansell™ Sol-Vex™ Nitrile Gloves
Resistant to chemicals—will not weaken or swell in many solvents, caustics or animal fats. Ansell™ Sol-Vex™ Nitrile Gloves are ideal for numerous applications involving chemical processing.
Manufacturer: Ansell™ 37185 7 117298
 View more versions of this product
Catalog No. 11-391-140A
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Glove Cuff Style Straight
    For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
    Length (English) 18 in.
    Length (Metric) 45.7cm
    Lining None
    Material Nitrile
    Palm Type Contoured
    Size 7
    Thickness 22mil
    Color Green
  • Description & Specifications
Related Products
Product Content Feedback