Ansell™ Sol-Vex™ Nitrile Gloves
Resistant to chemicals—will not weaken or swell in many solvents, caustics or animal fats. Ansell™ Sol-Vex™ Nitrile Gloves are ideal for numerous applications involving chemical processing.
Manufacturer: Ansell™ 37165 10 117210
 View more versions of this product
Catalog No. 11-391-138D
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Glove Cuff Style Straight
    For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
    Length (English) 15 in.
    Length (Metric) 38.1cm
    Material Nitrile
    Palm Type Contoured
    Size 10
    Thickness 22mil
    Color Green
    Lining None
  • Description & Specifications
Related Products
Product Content Feedback