Ansell™ STD™ Vinyl-Impregnated Gloves

Exceptionally flexible

Manufacturer: Ansell™ 01114 10 201002

 View more versions of this product

Catalog No. 19-014657

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Cleaning Procedure Machine washable
    Glove Cuff Style Slip-on
    Glove Cuff Color Brown
    For Use With (Application) assembling, inspecting and packing small or delicate parts
    Length (English) 8.66 to 9.25 in.
    Length (Metric) 22 to 23.5cm
    Material Vinyl
    Liner Material Cotton
    Size 10
    Color Brown

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit