Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ TouchNTuff™ Nonsterile Nitrile Gloves

Manufacturer:  Ansell™ 556079

 View more versions of this product

Catalog No. 19-823-522


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Textured fingertips
Ambidextrous
Yes
6.5,7,Small
8cm
CE
Green
11.81 in.
Beaded/Rolled
Highest protection for a wide range of chemicals than any other single use Nitrile glove
Nitrile
3.14 in.
0.135mm
Powdered
Non-sterile
30cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit