Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ TouchNTuff™ Sterile Polyisoprene Gloves

Minimizes the risk of latex sensitization.

Manufacturer:  Ansell™ 384217

 View more versions of this product

Catalog No. 19-823-513


  Check Availability
Add to cart

Specifications

Description

  • Ergonomically-designed hand specific formers enhance fit and comfort
  • Made from latex-free polyisoprene; does not contain natural rubber latex
  • Low allergenicity; minimizes type I allergy for latex
  • SureFit Technology maintains glove securely around arm, avoiding cuff roll down
  • Anti-slip finish helps prevent slippage and provides grip
Description & Specifications

Specifications

12.2 x 3.5 in. (310 x 89mm)
Polyisoprene
Yes
Polyisoprene
3.5 in.
8.6mil
Powder-free
Sterile
310mm
Yes
Beaded/Rolled
Ideal for analytical measurements, blending of liquids, filtration processes, pelletizing and tablet manufacturing, tablet press preparation and operation
7
89mm
ASTM D 3767, ASTM D 5151, ASTM D 7102, EN 374-2, EN 388 0493, EN 420, EN 421, IEST-RP-CC005.2-7
White
12.2 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit