Learn More
Ansell™ TRELLCHEM™ ACT Coveralls with Bayonet glove ring system
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
Requires
Onguard Hazmax™ boots and SCBA units by Scott and MSA
- Made of techinical barrier fabric with a fluoro-based barrier film
- Non-encapsulating design (Level B/TypeT) with mask seal and without hump, self-contained breathing apparatus (SCBA) worn outside the suit
- Elastic waist and shoulder paddings for added comfort
- Attached hood with 3D shaped butyl rubber mask seal
- long zipper positioned diagonally over the chest, closing down-wards for added safety. The zipper is protected by a double splash guard(flap)
- Bayonet glove ring system, which offers quick and simple glove replacement
- Equipped with sewn-in sock/bootie in suit material
- Seams are stitched with aramid thread sealed by a welded-on barrier strip, seamless barrier and optimal safety
- Available in XS to XXXL sizes
- Black in color
Optional Features:
- bullets1 (begin)External pockets on chest, waist and/or hip
- Paddings on knees and elbows for improved comfort and safety
- Reflective markings on sleeves and legs for high visibilitybullets1 (end)
- Law enforcement
- First responders
- Hazmat teams
- “Hot Zone”search and rescue
- Explosive Ordnance Disposal (EOD)
- Clandestine lab entry, drug raids
- Technical rescue in contaminated environment
- Decontamination
- Tactical operations: Critical infrastucture protection, Quick response forces
- CBRNE response
- Civil Support Teams: USAR teams, Triage, Medical operations
Compliance
American standard NFPA1994 (class2)
Specifications
Size | X-Large |
Material | Technical Barrier Fabric |
Color | Black |