Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ TRELLCHEM™ ACT Coveralls with Bayonet glove ring system

Catalog Number p-5237566
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Onguard Hazmax™ boots and SCBA units by Scott and MSA

Specially designed for protection in CBRN terrorism incidents and IDLH

  • Made of techinical barrier fabric with a fluoro-based barrier film
  • Non-encapsulating design (Level B/TypeT) with mask seal and without hump, self-contained breathing apparatus (SCBA) worn outside the suit
  • Elastic waist and shoulder paddings for added comfort
  • Attached hood with 3D shaped butyl rubber mask seal
  • long zipper positioned diagonally over the chest, closing down-wards for added safety. The zipper is protected by a double splash guard(flap)
  • Bayonet glove ring system, which offers quick and simple glove replacement
  • Equipped with sewn-in sock/bootie in suit material
  • Seams are stitched with aramid thread sealed by a welded-on barrier strip, seamless barrier and optimal safety
  • Available in XS to XXXL sizes
  • Black in color
Optional Features:
  • bullets1 (begin)External pockets on chest, waist and/or hip
  • Paddings on knees and elbows for improved comfort and safety
  • Reflective markings on sleeves and legs for high visibilitybullets1 (end)
Applications:
  • Law enforcement
  • First responders
  • Hazmat teams
  • “Hot Zone”search and rescue
  • Explosive Ordnance Disposal (EOD)
  • Clandestine lab entry, drug raids
  • Technical rescue in contaminated environment
  • Decontamination
  • Tactical operations: Critical infrastucture protection, Quick response forces
  • CBRNE response
  • Civil Support Teams: USAR teams, Triage, Medical operations

Compliance

American standard NFPA1994 (class2)

Size Small
Material Technical Barrier Fabric
Color Black
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit