Ansell™ Unsupported Neoprene Gloves
Embossed finish for better grip
Manufacturer: Ansell™ 29865 8 116312
 View more versions of this product
Catalog No. 11-391-130B
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600
    Glove Cuff Style Straight
    For Use With (Application) bulk chemical handling and transport, cleaning and de-greasing, handling Alkalies and Salts, handling Oils, Acids, Caustics and Alcohols, handling specialty chemicals
    Size 8
    Thickness 18mil
    Color Black
    Length (English) 13 in.
    Length (Metric) 33cm
    Lining Flock
    Material Neoprene
    Coating Material Sand patch
  • Description & Specifications
Related Products
Product Content Feedback