Learn More
Ansell™ Unsupported Neoprene Gloves
Embossed finish for better grip
Manufacturer:  Ansell™ 29865 9 116313
 View more versions of this product
Catalog No. 11-391-130C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Sand patch
bulk chemical handling and transport, cleaning and de-greasing, handling Alkalies and Salts, handling Oils, Acids, Caustics and Alcohols, handling specialty chemicals
Neoprene
18mil
Black
33cm
Straight
Flock
9
21 CFR 177-2600
13 in.