Ansell™ Unsupported Neoprene Gloves
Embossed finish for better grip
Manufacturer: Ansell™ 29865 10 116314
 View more versions of this product
Catalog No. 11-391-130D
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600
    Glove Cuff Style Straight
    For Use With (Application) bulk chemical handling and transport, cleaning and de-greasing, handling Alkalies and Salts, handling Oils, Acids, Caustics and Alcohols, handling specialty chemicals
    Material Neoprene
    Size 10
    Thickness 18mil
    Coating Material Sand patch
    Color Black
    Length (English) 13 in.
    Length (Metric) 33cm
    Lining Flock
  • Description & Specifications
Related Products
Product Content Feedback