Ansell™ Unsupported Neoprene Gloves
Embossed finish for better grip
Manufacturer: Ansell™ 29845 77.5 116237
 View more versions of this product
Catalog No. 11-391-129A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Certifications 21 CFR 177-2600
    Glove Cuff Style Straight
    For Use With (Application) bulk chemical handling and transport, Oil refining and Petrochemicals, cleaning and de-greasing, Detergents, handling equipment and abrasive materials coated with Oil, Grease, Acids and Solvents
    Length (Metric) 30.48cm
    Coating Material Sand patch
    Length (English) 12 in.
    Lining Unlined
    Material Neoprene
    Size 7
    Thickness 17mil
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback