Ansell™ Unsupported Neoprene Gloves
Embossed finish for better grip
Manufacturer: Ansell™ 29845 10 116243
 View more versions of this product
Catalog No. 11-391-129D
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Certifications 21 CFR 177-2600
    Glove Cuff Style Straight
    For Use With (Application) bulk chemical handling and transport, cleaning and de-greasing, handling Alkalies and Salts, handling Oils, Acids, Caustics and Alcohols, handling specialty chemicals
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Lining Unlined
    Material Neoprene
    Size 10
    Thickness 17mil
    Coating Material Sand patch
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback