Ansell™ Vantage™ Cut-Resistant Gloves
Superior cut protection, abrasion resistance and comfort
Manufacturer: Ansell™ 70761 9 245710
 View more versions of this product
Catalog No. 19-920-1772
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Knit
    For Use With (Application) appliance manufacturing, automotive, general carpentry, general industry, Glass, masonry/tile, Metal fabrication, Metalworking, mining, plumbers, textiles
    Material Kevlar™ Fiber
    Size 9
    Weight Medium weight
    Color Green
    Cut Resistance Rating Level 4
  • Description & Specifications
Related Products
Product Content Feedback