Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell VersaTouch™ 92-220 Nitrile Disposable Food Processing Gloves

Thin, disposable nitrile gloves with enhanced fingertip flexibility and overall comfort for safe food processing

Manufacturer:  Ansell 92220100

 View more versions of this product

Catalog No. 19-148-528



Add to cart
This item is not returnable. View return policy

Description

Description

  • Offers better grip in dry, wet and fatty food handling conditions
  • Textured fingertips provide good comfort and tactility for sensitive, precise handling
  • Latex-free with no risk of Type 1 allergies
Specifications

Specifications

White
10
9.4 in.
Category III; EN 420:2003 + A1:2009
Yes
3.9 mil
1.5
3.9 mil
2 mil
Nitrile
Powdered
240 mm
FDA21 CFR 177-2600
Yes
0.10 mm
Industrial
0.10 mm
0.05 mm
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit