Learn More
Ansell™ AlphaTec™ 56-910 Vinyl Closed-Back Aprons With Snap-on Ties
Catalog Number 17987116d
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
With heat-sealed elastic cuffs
  • Neck ties, 15 in. long (38cm); waist ties, 29 in. long (74cm)
  • 8mil thick aprons have electronically welded vinyl binding strip that covers all edges for smoother finish
  • 6mil thick aprons have electronically welded binding strip around neck edge only; other edges are unhemmed
  • Available with snap-on nylon ties
  • Blue
Order Info
Can be cleaned to ISO Class 5 (Class 100) cleanroom standards and shipped in heat-sealed bags; call your Fisher Safety Customer Service Representative at 1-800-772-6733 for details.
Compliance
Permeation tested in accordance with MIL-STD 282 (using various chemical warfare agents) and ASTM™ F739 (using ASTM F1001 test battery).
Size X-Large
Material Vinyl
Color Blue
Closure Type Snap-on Ties
Cuff Style Elastic
Thickness 8 mil