Learn More
Ansell™ Winter Monkey Grip™ 23-173 Series PVC Heavy Duty Gloves
Keeps hands warm, dry and protected from abrasions. Vinyl coating bonded to a needled nonwoven material.
Manufacturer:  Ansell™ 23152 10 204827
 View more versions of this product
Catalog No. 19-122-698
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ideal for work in construction, cold storage facilities, winter transportation, and outdoor jobs in cold weather. Fully coated with gauntlet, or safety cuff.
  • Resist chemicals and abrasion
  • Allow excellent grip for easy, safe handling of wet and dry materials
  • Foam insulation locks out cold, seals in warmth
  • Size 10
Compliance
Meets ANSI A1 (cut level) and ANSI 3 (abrasion); EN 388, EN 511, and EN 1149 standards.
Specifications
Specifications
Smooth
construction, cold storage facilities, winter transportation and outdoor jobs in cold weather
10
Gauntlet
Vinyl
Orange