Learn More
Brady™ ANSI Sign: CONFINED SPACE. AUTHORIZED PERSONNEL ONLY.
Improve workplace safety with cold temperature permanent acrylic mounted signage. The Brady™ Confined Space ANSI Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$11.25 - $25.75
Specifications
Color Red/Black on White
Description Polyester with polyester overlaminate
Message CONFINED SPACE. AUTHORIZED PERSONNEL ONLY.
Quantity 1 Ea.
Shape Rectangular
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-107-805
View Documents
Brady™
143656
10 x 7 in. Each for $11.25
Add to cart
19-107-806
View Documents
Brady™
143657
14 x 10 in. Each for $25.75
Add to cart
Description
Description
  • Polyester construction is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Strong adhesive back for indoor or outdoor use
Specifications
Specifications
Red/Black on White
CONFINED SPACE. AUTHORIZED PERSONNEL ONLY.
Rectangular
Polyester
Polyester with polyester overlaminate
1 Ea.
0.01 in.