Learn More
Brady™ ANSI Sign: CONFINED SPACE LOCK OUT AND TAG OUT REQUIRED BEFORE ENTRY.
Improve workplace safety with non-adhesive mounted signage. The Brady™ Confined Space ANSI Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
Manufacturer:  Brady™ 143694
 View more versions of this product
Catalog No. 19-107-840
Add to cart
Description
Description
  • Abrasion-resistant and chemical-resistant
  • Brady™ signs feature bright colors, bold text, and intuitive pictographs
  • Highly visible and easily understood
  • Corner holes provide easy mounting solutions in a variety of environments
Specifications
Specifications
Safety
Not laminated
1/Sign
10 in. x 7 in. x 0.035 in.
7 in.
CONFINED SPACE LOCK OUT AND TAG OUT REQUIRED BEFORE ENTRY
0.035 in.
Black/red on white
Aluminum
Rectangle
Corner holes
230°F
-40°F
10 in.