Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ANSI Sign: SLIPPERY WHEN WET. FALLS MAY RESULT IN INJURY.

Improve workplace safety with non-adhesive mounted signage. The Brady™ Slippery When Wet ANSI Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

Manufacturer:  Brady™ 144756

 View more versions of this product

Catalog No. 19-112-859


  Check Availability
Add to cart

Specifications

Description

  • Abrasion-resistant and chemical-resistant
  • Brady™ signs feature bright colors, bold text, and intuitive pictographs
  • Highly visible and easily understood
  • Corner holes provide easy mounting solutions in a variety of environments
Description & Specifications

Specifications

Black/yellow on white
10 in.
SLIPPERY WHEN WET. FALLS MAY RESULT IN INJURY.
Rectangle
-40°F to 176°F
14 in.
Not laminated
Polystyrene
1/Sign
14 in. x 10 in. x 0.0591 in.
0.0591 in.
Corner holes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit