Learn More
PIP™ ANSI Type R Class 2 Two-Tone X-Back Breakaway Mesh Vest
Type R/Class 2 apparel is necessary for workers exposed to traffic traveling over 25 mph and those who work against complex backgrounds. Ideal for railway workers, construction, municipalities, shipyards, or anywhere that hi-vis apparel is required.
$12.62
Specifications
Certifications/Compliance ANSI/ISEA 107-2015 Class 2 Type R
Closure Type Hook & Loop
Color Hi-Vis Yellow
Construction 5 Point Breakaway/X-Back
Material Polyester Mesh
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-046-493
View Documents
PIP™
3020211LY/S
Small
Each for $12.62
Add to cart
19-046-494
View Documents
PIP™
3020211LY/M
Medium
Each for $12.62
Add to cart
19-046-495
View Documents
PIP™
3020211LY/L
Large
Each for $12.62
Add to cart
19-046-496
View Documents
PIP™
3020211LY/XL
X-Large
Each for $12.62
Add to cart
19-046-497
View Documents
PIP™
3020211LY/2X
2X-Large
Each for $12.62
Add to cart
19-046-499
View Documents
PIP™
3020211LY/3X
3X-Large
Each for $12.62
Add to cart
19-046-500
View Documents
PIP™
3020211LY/4X
4X-Large
Each for $12.62
Add to cart
19-046-501
View Documents
PIP™
3020211LY/5X
5X-Large
Each for $12.62
Add to cart
19-046-502
View Documents
PIP™
3020211LY/6X
6X-Large
Each for $12.62
Add to cart
Description
Description
  • Breathable, 5 point breakaway polyester mesh
  • 2 in. two-tone reflective tape for optimum visibility
  • Hook & Loop closure
  • 2 external lower flap pockets
  • 2 upper chest pockets, including a radio pocket
  • 2 mic tabs
Specifications
Specifications
ANSI/ISEA 107-2015 Class 2 Type R
Hi-Vis Yellow
Polyester Mesh
4
Hook & Loop
5 Point Breakaway/X-Back
Construction, Municipalities, Shipyards, and Railroads