Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ANSI Type R Class 2 X-Back Breakaway Mesh Vest

Type R/Class 2 apparel is necessary for workers exposed to traffic traveling over 25 mph and those who work against complex backgrounds. Ideal for railway workers, construction, municipalities, shipyards, or anywhere that hi-vis apparel is required.

$11.02

Specifications

Certifications/Compliance ANSI/ISEA 107-2015 Class 2 Type R
Closure Type Hook & Loop
Color Hi-Vis Orange
Construction 5 Point Breakaway/X-Back
For Use With (Application) Construction, Municipalities, Shipyards, and Railroads
View More Specs

Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-046-486
View Documents
PIP™
3020210OR/S
Small
Each for $11.02
Add to cart
 
19-046-487
View Documents
PIP™
3020210OR/M
Medium
Each for $11.02
Add to cart
 
19-046-488
View Documents
PIP™
3020210OR/L
Large
Each for $11.02
Add to cart
 
19-046-489
View Documents
PIP™
3020210OR/XL
X-Large
Each for $11.02
Add to cart
 
19-046-490
View Documents
PIP™
3020210OR/2X
2X-Large
Each for $11.02
Add to cart
 
19-046-491
View Documents
PIP™
3020210OR/3X
3X-Large
Each for $11.02
Add to cart
 
19-046-492
View Documents
PIP™
3020210OR/4X
4X-Large
Each for $11.02
Add to cart
 
Description

Description

  • Breathable, breakaway polyester mesh
  • 5 point breakaway
  • 2 in. silver reflective tape
  • Hook & Loop closure
  • 2 external lower flap pockets
  • 1 upper chest pocket and mic tab
Specifications

Specifications

ANSI/ISEA 107-2015 Class 2 Type R
Hi-Vis Orange
Construction, Municipalities, Shipyards, and Railroads
3
Hook & Loop
5 Point Breakaway/X-Back
Polyester Mesh
Level 2 Silver Reflective (2 in.)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit