Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansul™ Ansulite™ 6% (AFC-5) AFFF Foam Concentrate

For use on Class A fires and Class B hydrocarbon fuel fires having low water solubility

Manufacturer:  Ansul™ 68118

 View more versions of this product

Catalog No. 19-806-184

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Premium AFFF Concentrate is formulated from specialty fluorochemical and hydrocarbon surfactants along with solvents. Intended for use as a 6% proportioned solution in fresh, salt, or hard water.

  • Offers excellent wetting characteristics
  • Suitable for use on fuel fires such as crude oils, gasoline, diesel fuels, and aviation fuels
  • Can be used both with aspirating and non-aspirating discharge devices

Certifications

UL listed

Compliance

Meets MIL-F-24385/QPL-24385-31.

Specifications

Specifications

1 in.
2.54cm
Fluorochemical and hydrocarbon surfactants, a high molecular weight polymer and solvent
Ansulite 6% (AFC-5) AFFF Foam
Fuel fires such as crude Oils, gasoline, diesel fuels and aviation fuels
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit