Learn More
Ansul™ Ansulite™ 6% (AFC-5) AFFF Foam Concentrate
For use on Class A fires and Class B hydrocarbon fuel fires having low water solubility
Specifications
Formulation Fluorochemical and hydrocarbon surfactants, a high molecular weight polymer and solvent
Product Type Ansulite 6% (AFC-5) AFFF Foam
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-806-184
View Documents Promotion Details
Ansul™
68118
Bulk Shipment (per gallon), Ansulite 6% (AFC-5) AFFF Bulk Shipment (per gallon), Ansulite 6% (AFC-5) AFFF This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Premium AFFF Concentrate is formulated from specialty fluorochemical and hydrocarbon surfactants along with solvents. Intended for use as a 6% proportioned solution in fresh, salt, or hard water.
  • Offers excellent wetting characteristics
  • Suitable for use on fuel fires such as crude oils, gasoline, diesel fuels, and aviation fuels
  • Can be used both with aspirating and non-aspirating discharge devices
Certifications
UL listed
Compliance
Meets MIL-F-24385/QPL-24385-31.
Specifications
Specifications
Fluorochemical and hydrocarbon surfactants, a high molecular weight polymer and solvent
Ansulite 6% (AFC-5) AFFF Foam