ANSUL™ Ansulite™ 6% (AFC-5) AFFF Foam Concentrate

For use on Class A fires and Class B hydrocarbon fuel fires having low water solubility

Manufacturer: Ansul Fire 68120

 View more versions of this product

Catalog No. 19-120-3527

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 5 gal.
    Capacity 5 gal., 18.9L
    For Use With (Application) Class B hydrocarbon fuel fires such as crude Oils, gasoline, diesel fuels and aviation fuels
    Formulation Fluorochemical and hydrocarbon surfactants, a high molecular weight polymer and solvent
    Product Type Ansulite 6% (AFC-5) AFFF Foam
    Capacity (Metric) 18.9L

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit