Learn More
Ansul™ Ansulite Premium Foams
Manufacturer:  Ansul™ AFC5
Catalog No. 19-500-158
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
5 gal.
5 gal., 18927.1mL
Specialty fluorochemical/hydrocarbon surfactants with solvents
18927.1mL
Ideal for suppressing fires involving petroleum-based products, LNG, rubber, and flammable and combustible liquids such as diesel fuel, crude oil gasoline and ethanol
Ansulite Premium Foam