ANSUL™ Bulk SPILL-X™ Agents

For response to larger spills

Manufacturer: Ansul Fire 76255

 View more versions of this product

Catalog No. 17-200-1A

  Check Availability
Add to Cart

Alert

Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste material should comply with local, state, and federal regulations.


    Specifications

    Color Red
    Density 0.83g/cc
    Product Type Spill-X-A Agent
    Base Magnesium Oxide
    For Use With (Application) To solidify spills of acids
    For Use With (Equipment) For Spill Gun™ applicator
    Includes Detailed instructions for responding to a spill
    Size (English) 5 gal.
    Size (Metric) 19L
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -54° to 49°C
    Weight (English) 50 lb.
    Weight (Metric) 22.7kg

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit