Learn More
ANSUL™ Bulk SPILL-X™ Agents
For response to larger spills
Manufacturer:  Ansul Fire 76258
 View more versions of this product
Catalog No. 17-200-1B
  Check Availability
Add to cart
Alert:
Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste material should comply with local, state, and federal regulations.
Specifications Description & Specifications
Specifications
Carbonaceous
0.48g/cc
For Spill Gun™ applicator
5 gal.
-65° to 120°F
Spill-X-S Agent
7.3kg
Black
To solidify spills of solvents
Detailed instructions for responding to a spill
19L
-54° to 49°C
16 lb.
Accessories