Learn More
Ansul™ Dry Chemicals
Dry chemicals are available to fight Class A, B, C, or D fires
Supplier:  Ansul™ 9328
 View more versions of this product
Catalog No. 18-999-1874
Add to cart
Description
Description
Never mix bicarbonate-based and phosphate-based dry chemicals. A chemical reaction that is harmful to the extinguisher will occur. Foray™ is free flowing, water repellent, nonabrasive; and produces no toxic effects. Other additives to Met-L-X™ render it free-flowing and cause heat caking or crusting.
Foray
▪    For Class A, B or C fires
▪    Composed of a monoammonium phosphate base
▪    Useful on combination fires and on Class A fires at temperatures that freeze water; applications include heavy equipment used in the logging industry
▪    May produce mild corrosion after fire, making prompt cleanup necessary; if agent cannot be readily swept up, warm water may be used as a solvent
Met-L-X
▪    For Class D (sodium, potassium, sodium-potassium alloy or magnesium) fires
▪    Composed of a salt base plus a polymer for sealing
▪    Controls and sometimes extinguishes small fires on zirconium and titanium
▪    UL listed
Specifications
Specifications
Met-L-X
Class D (Sodium, Potassium, Sodium Potassium Alloy or Magnesium) fires
22.7 kg
Composed of a salt base plus a polymer for sealing
50 lb.