Learn More
ANSUL™ Dry Chemicals
Dry chemicals are available to fight Class A, B, C, or D fires
Manufacturer:  Ansul Fire 53080
 View more versions of this product
Catalog No. 19-029651
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Foray
Composed of a salt base plus a polymer for sealing
20.4kg
Class A, B, C fires
45 lb.