Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansul™ Fire Sentry™ 5 Multipurpose Portable Extinguishers

For both industrial and commercial fire-protection needs

Specifications

Product Type Fire extinguisher
Discharge Time 14 sec.
Includes Vehicle mounting bracket
Material Steel
Range 14 ft.
View More Specs

Products 2
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Model Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Model Price Quantity & Availability  
19-806-189
View Documents
Ansul™
SY0516
18 lb., 8.2kg 4.8 x 7.35 x 16.85 in. (12.19 x 18.66 x 42.7cm) SY-0516 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-065506
View Documents
Ansul™
429001
5 lb., 2.26kg 4.4 x 8.5 x 17 in. (11.3 x 21.6 x 43.1cm) A05 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Sentry dry-chemical extinguishers are designed for protection of light and ordinary hazards.

  • Competitive fire ratings
  • Simple operation and maintenance
  • Compact and portable
  • Tough, glare-resistant, one-piece mylar nameplates
  • Rechargeable
  • Manufactured of durable, high-quality materials
  • Large and easy-to-read pressure gauge
  • Fits most wall-mounted storage cabinets

  • Agent Capacity: 2.27kg (5 lb.)
  • Coast Guard Classification: Type A, Size II; Type B, C, Size I
  • Discharge time: 14 seconds
  • Charged weight (with Agent): 4.15kg (9.15 lb.)
  • Max. Effective Range: 4.3m (14 ft.)

  • Wall bracket available (19-029-577, 19-080-569)

Compliance

ANSI/UL 299 and 711; DOT and USCG approved with proper bracket

Warranty and Services

Six-year, limited

Specifications

Specifications

Fire extinguisher
Vehicle mounting bracket
14 ft.
5 lb.
14 sec.
Steel
3A:40B:C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit