Ansul™ Fire Sentry™ 5 Multipurpose Portable Extinguishers

For both industrial and commercial fire-protection needs

Manufacturer: Ansul Fire SY0516

 View more versions of this product

Catalog No. 19-806-189

  Check Availability
Add to Cart

    Specifications

    Capacity 18 lb., 8.2kg
    Width (English) 7.35 in.
    Capacity (Metric) 8.2kg
    Product Type Fire extinguisher
    Dimensions (L x W x H) 4.8 x 7.35 x 16.85 in. (12.19 x 18.66 x 42.7cm)
    Discharge Time 14 sec.
    Height (Metric) 42.7cm
    Includes Vehicle mounting bracket
    Length (English) 4.8 in.
    Length (Metric) 12.19cm
    Material Steel
    Range 14 ft.
    UL Rating 3A:40B:C
    Weight (English) 5 lb.
    Width (Metric) 18.66cm
    Capacity (English) 18 lb.
    Height (English) 16.85 in.
    Model SY-0516
    Reagent Type Dry chemical

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit