Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansul™ Red Line™ Wheeled Fire Extinguisher

Provides protection to hazardous areas where the potential for a large fire exists

Manufacturer:  Ansul™ 53874

Catalog No. 19-065-591

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

UL rating: 320-B:C. Contains 125 lb. (56.7kg) Purple-K dry chemical extinguishing agent. Charged weight: 329 lb. (149.2kg). Dimensions: 36D x 24W x 45 1/2 in.H (91 x 61 x 116cm). Hose length: 100 ft. (30.4m).

  • Requires only one trained person for operation
  • Occupies minimal space
  • Requires no outside pressure source to completely expel extinguishing agent from welded steel tank

Specifications

Specifications

125 lb., 56.6kg
48 sec.
115.57cm
91.44cm
CR-I-K-150-D-1
Purple K
40° to 49°C
24 in.
125 lb.
Wheeled red line extinguisher
36 x 24 x 45.5 in. (91.44 x 61 x 115.57cm)
45.5 in.
36 in.
Steel
35 ft.
-40° to 120°F
320B:C
61cm
56.6kg
Safety
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit