ANSUL™ Sentry™ 20 CO2 Extinguisher

Manufacturer: Ansul Fire CD20A1

 View more versions of this product

Catalog No. 18-771D

  Check Availability
Add to Cart

    Specifications

    Capacity 20 lb., 9.2kg
    Dimensions (L x W x H) 8.1 x 12 x 28 in. (20.6 x 30.5 x 71cm)
    Discharge Time 13 sec.
    Includes Wall mounted hanger
    Length (English) 8.1 in.
    Length (Metric) Hose 91.4cm
    Length (Metric) 20.6cm
    Material Aluminum
    Model CD20A-1
    Temperature Range (English) -40° to 120°F
    Width (English) 12 in.
    Width (Metric) 30.5cm
    Capacity (Metric) 9.2kg
    Product Type Hand portable fire extinguisher
    Height (English) 28 in.
    Height (Metric) 71cm
    Length (English) Hose 36 in.
    Range 6 ft.
    Reagent Type Carbon Dioxide
    Temperature Range (Metric) 40° to 49°C
    UL Rating 10B:C
    Capacity (English) 20 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit