Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansul™ Sentry™ 20 CO2 Extinguisher

Manufacturer:  Ansul™ 431572

 View more versions of this product

Catalog No. 19-806-182

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

20 lb., 9.2kg
13 sec.
30.2cm
7.9 in.
20.1cm
CD20-1
Carbon Dioxide
40° to 49°C
11.8 in.
20 lb.
Fire extinguisher
7.9 x 11.8 x 28 in. (20.1 x 30.2 x 30.2cm)
28 in.
36 in.
91.4cm
Steel
6 ft.
-40° to 120°F
10B:C
30.2cm
9.2kg
Safety
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit