Learn More
ANSUL™ SPILL-X™ Agents Spill Treatment Kits
Mount easily on a wall yet remove quickly
Manufacturer:  Ansul Fire 78776
 View more versions of this product
Catalog No. 17-987-144B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For acid
6 agents in shaker bottles, instructions, guide, goggles, gloves, disposal bags, scraper, pH test papers, pick up pans, green suitcase
Easily removable wall mount
Single Agent Kit
Accessories